ENAH5802E954 Enamine

5-methanesulfonyl-1-methyl-1H,4H,5H,6H,7H-pyrazolo[4,3-c]pyridine-4-carboxylic acid

  • MDL number MFCD32198252
  • Cas Number : 2375259-33-7
  • Molecular Weight : 259.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-methyl-5-(methylsulfonyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-4-carboxylic acid

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6504174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6504174
Availability details for
Quantity
  • Availability
  • Add to Cart