ENAH5802E956 Enamine

2-{4-[(4-fluorophenyl)methoxy]phenyl}acetic acid

  • MDL number MFCD12621888
  • Molecular Weight : 260.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name {4-[(4-fluorobenzyl)oxy]phenyl}acetic acid
Inchi Code 1S/C15H13FO3/c16-13-5-1-12(2-6-13)10-19-14-7-3-11(4-8-14)9-15(17)18/h1-8H,9-10H2,(H,17,18)
Inchi Key FLXSYWHTSCMPAZ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7430747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430747
Availability details for
Quantity
  • Availability
  • Add to Cart