ENAH5802E957 Enamine

2-acetamido-3-(7-methyl-1H-indol-3-yl)propanoic acid

  • MDL number MFCD22384361
  • Cas Number : 71953-92-9
  • Molecular Weight : 260.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-369991

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-369991
Availability details for
Quantity
  • Availability
  • Add to Cart