ENAH5802E958 Enamine

3-phenyl-2-(2,2,2-trifluoroacetamido)propanoic acid

  • MDL number MFCD00441734
  • Cas Number : 2728-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443931
Availability details for
Quantity
  • Availability
  • Add to Cart