ENAH5802E966 Enamine

(2S)-3-(4-hydroxyphenyl)-2-[2-(methylsulfanyl)acetamido]propanoic acid

  • MDL number MFCD16378377
  • Cas Number : 1189191-19-2
  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S)-3-(4-hydroxyphenyl)-2-{[(methylsulfanyl)acetyl]amino}propanoic acid
Inchi Code 1S/C12H15NO4S/c1-18-7-11(15)13-10(12(16)17)6-8-2-4-9(14)5-3-8/h2-5,10,14H,6-7H2,1H3,(H,13,15)(H,16,17)/t10-/m0/s1
Inchi Key WWCIZXHEXDWJEZ-JTQLQIEISA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7431686

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7431686
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart