ENAH5802E968 Enamine

(1s,4s)-1-[(1H-1,3-benzodiazol-2-yl)methyl]-4-methylcyclohexane-1-carboxylic acid hydrochloride

  • Cas Number : 2375269-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7444202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444202
Availability details for
Quantity
  • Availability
  • Add to Cart