ENAH5802E96C Enamine

(2S)-2-[(furan-3-yl)formamido]-3-(4-hydroxyphenyl)propanoic acid

  • MDL number MFCD32068905
  • Cas Number : 1102895-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434497
Availability details for
Quantity
  • Availability
  • Add to Cart