ENAH5802E96F Enamine

lithium(1+) ion 6-[(tert-butoxy)carbonyl]-2-oxa-6-azaspiro[3.4]octane-8-carboxylate

  • Cas Number : 2344685-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Product Link https://www.enaminestore.com/catalog/EN300-7354435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7354435
Availability details for
Quantity
  • Availability
  • Add to Cart