ENAH5802E976 Enamine

(2S)-3-(4-hydroxyphenyl)-2-[2-(propan-2-yloxy)acetamido]propanoic acid

  • MDL number MFCD32066282
  • Cas Number : 1922901-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7434418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434418
Availability details for
Quantity
  • Availability
  • Add to Cart