ENAH5802E977 Enamine

diphenyl-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD13187291
  • Cas Number : 425392-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-138
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7438408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438408
Availability details for
Quantity
  • Availability
  • Add to Cart