ENAH5802E97B Enamine

3-(furan-2-yl)-2-(5-phenyl-1H-1,2,3,4-tetrazol-1-yl)prop-2-enoic acid

  • Cas Number : 19747-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-09248

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09248
Availability details for
Quantity
  • Availability
  • Add to Cart