ENAH5802E97F Enamine

5-{[2-methoxy-4-(prop-2-en-1-yl)phenoxy]methyl}furan-2-carboxylic acid

  • MDL number MFCD02090872
  • Molecular Weight : 288.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-[(4-allyl-2-methoxyphenoxy)methyl]-2-furoic acid
Inchi Code 1S/C16H16O5/c1-3-4-11-5-7-13(15(9-11)19-2)20-10-12-6-8-14(21-12)16(17)18/h3,5-9H,1,4,10H2,2H3,(H,17,18)
Inchi Key NYGXLNNKIZBLJY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432840

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432840
Availability details for
Quantity
  • Availability
  • Add to Cart