ENAH5802E981 Enamine

(2S)-3-(4-hydroxyphenyl)-2-[(thiophen-3-yl)formamido]propanoic acid

  • Cas Number : 1101186-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7431868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431868
Availability details for
Quantity
  • Availability
  • Add to Cart