ENAH5802E983 Enamine

5-[butyl(methyl)amino]-2-(2-methylpropanamido)benzoic acid

  • MDL number MFCD27184730
  • Cas Number : 1243028-14-9
  • Molecular Weight : 292.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(butyl(methyl)amino)-2-isobutyramidobenzoic acid
Inchi Code 1S/C16H24N2O3/c1-5-6-9-18(4)12-7-8-14(13(10-12)16(20)21)17-15(19)11(2)3/h7-8,10-11H,5-6,9H2,1-4H3,(H,17,19)(H,20,21)
Inchi Key RWKPGUAOUPTMGZ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7437410

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7437410
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart