ENAH5802E985 Enamine

1-benzyl-4-phenylpiperidine-4-carboxylic acid

  • MDL number MFCD00071791
  • Cas Number : 61886-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 285-286
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432353
Availability details for
Quantity
  • Availability
  • Add to Cart