ENAH5802E98C Enamine

2-benzyl-4-imino-5-methyl-1H,4H-thieno[2,3-d]pyrimidine-6-carboxylic acid

  • MDL number MFCD26143698
  • Cas Number : 924147-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26416
Availability details for
Quantity
  • Availability
  • Add to Cart