ENAH5802E98D Enamine

4,5-dibromo-3-methylthiophene-2-carboxylic acid

  • MDL number MFCD27930042
  • Cas Number : 854626-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-315191

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-315191
Availability details for
Quantity
  • Availability
  • Add to Cart