ENAH5802E991 Enamine

2-[2-(2,6-dioxopiperidin-3-yl)-1-oxo-2,3-dihydro-1H-isoindol-4-yl]acetic acid

  • Cas Number : 2375260-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6504629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6504629
Availability details for
Quantity
  • Availability
  • Add to Cart