ENAH5802E9A1 Enamine

3-(1,3-benzothiazol-2-yl)-4-(3-methoxyphenyl)but-3-enoic acid

  • Cas Number : 748786-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-00240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00240
Availability details for
Quantity
  • Availability
  • Add to Cart