ENAH5802E9A2 Enamine

3-(1,3-benzothiazol-2-yl)-4-(4-methoxyphenyl)but-3-enoic acid

  • Cas Number : 748786-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00238
Availability details for
Quantity
  • Availability
  • Add to Cart