ENAH5802E9B5 Enamine

2-[(1E)-2-phenylethenesulfonyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

  • MDL number MFCD03967080

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427139
Availability details for
Quantity
  • Availability
  • Add to Cart