ENAH5802E9B6 Enamine

1-[(tert-butoxy)carbonyl]-3-({[(tert-butoxy)carbonyl]amino}methyl)pyrrolidine-3-carboxylic acid

  • MDL number MFCD12408591
  • Cas Number : 1158759-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7356391

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7356391
Availability details for
Quantity
  • Availability
  • Add to Cart