ENAH5802E9CC Enamine

2-[(4-methoxyphenyl)methyl]-1-oxo-3-phenyl-octahydro-1H-isoindole-3a-carboxylic acid

  • MDL number MFCD32066315
  • Cas Number : 2361635-03-0
  • Molecular Weight : 379.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1726198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726198
Availability details for
Quantity
  • Availability
  • Add to Cart