ENAH5802E9CD Enamine

(3S)-6-{[(benzyloxy)carbonyl]amino}-3-{[(tert-butoxy)carbonyl]amino}hexanoic acid

  • MDL number MFCD23136909
  • Cas Number : 65581-30-8
  • Molecular Weight : 380.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7437730

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7437730
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart