ENAH5802EA3F Enamine

2,2-dimethyl-3-[(piperidin-4-yl)amino]propanoic acid dihydrochloride

  • MDL number MFCD32066040
  • Cas Number : 2361643-98-1
  • Molecular Weight : 273.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432311
Availability details for
Quantity
  • Availability
  • Add to Cart