ENAH5802EA46 Enamine

methyl 2,2-dimethyl-3-[(piperidin-4-yl)amino]propanoate

  • MDL number MFCD31924030
  • Cas Number : 1870131-28-4
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-4595620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4595620
Availability details for
Quantity
  • Availability
  • Add to Cart