ENAH5802EA68 Enamine

methyl 1,2-thiazole-3-carboxylate

  • MDL number MFCD18381997
  • Cas Number : 1001907-72-7
  • Molecular Weight : 143.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-5101351

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5101351
Availability details for
Quantity
  • Availability
  • Add to Cart