ENAH5802EA7B Enamine

methyl 4,4-difluoro-3,3-dimethylpentanoate

  • MDL number MFCD31924109
  • Cas Number : 2015624-53-8
  • Molecular Weight : 180.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6487851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6487851
Availability details for
Quantity
  • Availability
  • Add to Cart