ENAH5802EA7D Enamine

tert-butyl cyclohexanecarboxylate

  • MDL number MFCD11976277
  • Cas Number : 16537-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7426843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426843
Availability details for
Quantity
  • Availability
  • Add to Cart