ENAH5802EA80 Enamine

methyl 2-[(trifluoromethyl)sulfanyl]propanoate

  • MDL number MFCD32198446
  • Cas Number : 1621595-07-0
  • Molecular Weight : 188.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-((trifluoromethyl)thio)propanoate
Inchi Code 1S/C5H7F3O2S/c1-3(4(9)10-2)11-5(6,7)8/h3H,1-2H3
Inchi Key JGGKRHDACDANOV-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7443303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443303
Availability details for
Quantity
  • Availability
  • Add to Cart