ENAH5802EA82 Enamine

methyl 6-methylimidazo[1,5-a]pyridine-1-carboxylate

  • MDL number MFCD32068831
  • Cas Number : 2104923-68-2
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 6-methylimidazo[1,5-a]pyridine-1-carboxylate
Inchi Code 1S/C10H10N2O2/c1-7-3-4-8-9(10(13)14-2)11-6-12(8)5-7/h3-6H,1-2H3
Inchi Key CAXQMMOIWKYXEG-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7427612

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7427612
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart