ENAH5802EA84 Enamine

methyl 8-methylimidazo[1,5-a]pyridine-1-carboxylate

  • Cas Number : 2107688-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7427107

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427107
Availability details for
Quantity
  • Availability
  • Add to Cart