ENAH5802EA8C Enamine

methyl 5-tert-butylpyridine-3-carboxylate hydrochloride

  • MDL number MFCD31924364
  • Cas Number : 2344679-71-4
  • Molecular Weight : 229.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7355308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355308
Availability details for
Quantity
  • Availability
  • Add to Cart