ENAH5802EA8D Enamine

ethyl 5-cyano-3-methylthiophene-2-carboxylate

  • MDL number MFCD26387110
  • Cas Number : 1034981-21-9
  • Molecular Weight : 195.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-cyano-3-methylthiophene-2-carboxylate
Inchi Code 1S/C9H9NO2S/c1-3-12-9(11)8-6(2)4-7(5-10)13-8/h4H,3H2,1-2H3
Inchi Key HFXQOKWUVZAUKT-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7427697

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7427697
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart