ENAH5802EA8E Enamine

methyl 2-[(dimethyl-S-aminosulfonimidoyl)amino]acetate hydrochloride

  • MDL number MFCD32066086
  • Cas Number : 2376726-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432717

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432717
Availability details for
Quantity
  • Availability
  • Add to Cart