ENAH5802EA8F Enamine

methyl 6-methyl-5-nitropyridine-2-carboxylate

  • MDL number MFCD09260861
  • Cas Number : 64215-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135254
Availability details for
Quantity
  • Availability
  • Add to Cart