ENAH5802EA90 Enamine

methyl 4,6-dimethoxypyridine-3-carboxylate

  • MDL number MFCD31567723

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7434412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434412
Availability details for
Quantity
  • Availability
  • Add to Cart