ENAH5802EA92 Enamine

methyl 5-(1,1-difluoroethyl)pyridine-2-carboxylate

  • Cas Number : 2113847-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7444195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444195
Availability details for
Quantity
  • Availability
  • Add to Cart