ENAH5802EA9C Enamine

methyl 2-chloro-1,3-benzoxazole-7-carboxylate

  • MDL number MFCD20528219
  • Cas Number : 909854-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7356183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7356183
Availability details for
Quantity
  • Availability
  • Add to Cart