ENAH5802EA9E Enamine

methyl 3-[imino(methyl)oxo-lambda6-sulfanyl]benzoate

  • MDL number MFCD32069086
  • Cas Number : 1027730-86-4
  • Molecular Weight : 213.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-(amino-(methylene)sulfinyl)benzoate
Inchi Code 1S/C9H11NO3S/c1-13-9(11)7-4-3-5-8(6-7)14(2,10)12/h3-6H,2H2,1H3,(H2,10,12)
Inchi Key VLDPAPKSVIFFBE-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1078362

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1078362
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart