ENAH5802EA9F Enamine

methyl 3-(cyanomethyl)-1H-indole-4-carboxylate

  • MDL number MFCD09954799
  • Cas Number : 169546-86-5
  • Molecular Weight : 214.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7440054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440054
Availability details for
Quantity
  • Availability
  • Add to Cart