ENAH5802EAA2 Enamine

methyl 3-(2,2-dimethylthiomorpholin-4-yl)propanoate

  • MDL number MFCD32069419
  • Cas Number : 1596616-95-3
  • Molecular Weight : 217.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-(2,2-dimethylthiomorpholino)propanoate
Inchi Code 1S/C10H19NO2S/c1-10(2)8-11(6-7-14-10)5-4-9(12)13-3/h4-8H2,1-3H3
Inchi Key AODZGNVORZGUJT-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7436732

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7436732
pictograms GHS05,GHS07
hstatements H302,H312,H315,H318,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart