ENAH5802EAA5 Enamine

ethyl 2-(1-methyl-1H-indazol-3-yl)acetate

  • Cas Number : 919766-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7426386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426386
Availability details for
Quantity
  • Availability
  • Add to Cart