ENAH5802EAA9 Enamine

ethyl 2-(3,4-dihydro-2H-1-benzopyran-4-yl)acetate

  • MDL number MFCD11976262
  • Cas Number : 119304-96-0
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3,4-dihydro-2H-chromen-4-ylacetate
Inchi Code 1S/C13H16O3/c1-2-15-13(14)9-10-7-8-16-12-6-4-3-5-11(10)12/h3-6,10H,2,7-9H2,1H3
Inchi Key UKJHGKSYAFMJHL-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7434878

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7434878
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart