ENAH5802EAAA Enamine

methyl 1-[imino(methyl)oxo-lambda6-sulfanyl]piperidine-4-carboxylate hydrochloride

  • MDL number MFCD32197765
  • Cas Number : 2375271-24-0
  • Molecular Weight : 256.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 1-(amino-(methylene)sulfinyl)piperidine-4-carboxylate hydrochloride

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7439757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439757
Availability details for
Quantity
  • Availability
  • Add to Cart