ENAH5802EAB1 Enamine

methyl 3-(4-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylate

  • MDL number MFCD14156090
  • Molecular Weight : 235.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-(4-fluorophenyl)-5-methylisoxazole-4-carboxylate
Inchi Code 1S/C12H10FNO3/c1-7-10(12(15)16-2)11(14-17-7)8-3-5-9(13)6-4-8/h3-6H,1-2H3
Inchi Key JTXOFNWDTGMOHG-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7431309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431309
Availability details for
Quantity
  • Availability
  • Add to Cart