ENAH5802EAB2 Enamine

tert-butyl 6-cyclopropyl-5-fluoropyridazine-3-carboxylate

  • MDL number MFCD32198223
  • Cas Number : 2375261-53-1
  • Molecular Weight : 238.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 6-cyclopropyl-5-fluoropyridazine-3-carboxylate

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7434036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7434036
Availability details for
Quantity
  • Availability
  • Add to Cart