ENAH5802EAB3 Enamine

methyl 3-(2,4-difluorophenyl)-1,2-oxazole-5-carboxylate

  • MDL number MFCD32066481
  • Cas Number : 2361643-73-2
  • Molecular Weight : 239.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-(2,4-difluorophenyl)isoxazole-5-carboxylate
Inchi Code 1S/C11H7F2NO3/c1-16-11(15)10-5-9(14-17-10)7-3-2-6(12)4-8(7)13/h2-5H,1H3
Inchi Key VIEZPZNLHMCIRL-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432004
Availability details for
Quantity
  • Availability
  • Add to Cart