ENAH5802EAB5 Enamine

1,2-di-tert-butyl aziridine-1,2-dicarboxylate

  • MDL number MFCD32197978
  • Cas Number : 127700-62-3
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name di-tert-butyl aziridine-1,2-dicarboxylate
Inchi Code 1S/C12H21NO4/c1-11(2,3)16-9(14)8-7-13(8)10(15)17-12(4,5)6/h8H,7H2,1-6H3
Inchi Key HADPXKYCDSMWJQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7442834

Safety Information

signalword Danger
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P310,P312,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7442834
pictograms GHS05,GHS07
hstatements H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart