ENAH5802EAB7 Enamine

ethyl 3-oxa-7-thia-4-azatricyclo[6.4.0.0,2,6]dodeca-1(8),2(6),4,9,11-pentaene-5-carboxylate

  • MDL number MFCD31915245
  • Cas Number : 2105215-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7352360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7352360
Availability details for
Quantity
  • Availability
  • Add to Cart